missing translation for 'onlineSavingsMsg'
Learn More

1-(chloromethyl)-4-(trifluoromethoxy)benzene, 97%, Thermo Scientific™

Product Code. 10110712
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10110712 10 g Amber Glass Bottle 10g
10335302 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10110712 Supplier Thermo Scientific Maybridge Supplier No. TL00386EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 65796-00-1
Molecular Formula C8H6ClF3O
Molecular Weight (g/mol) 210.58
MDL Number MFCD00052326
InChI Key LBMKFQMJURUPKC-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy benzyl chloride, 1-chloromethyl-4-trifluoromethoxy benzene, 4-trifluoromethoxy benzylchloride, 4-trifluoromethoxybenzyl chloride, benzene, 1-chloromethyl-4-trifluoromethoxy, 4-chloromethyl phenoxy trifluoromethane, pubchem5804, acmc-209ns9, ksc623c1l, p-trifluoromethoxybenzyl chloride
PubChem CID 2777261
IUPAC Name 1-(chloromethyl)-4-(trifluoromethoxy)benzene
SMILES C1=CC(=CC=C1CCl)OC(F)(F)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH2ClC6H4OCF3
Quantity 10 g
Formula Weight 210.58
Percent Purity 97%
Chemical Name or Material 1-(Chloromethyl)-4-(trifluoromethoxy)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.