missing translation for 'onlineSavingsMsg'
Learn More

1-(Chloromethyl)-3,5-bis[[(1,1-dimethylethyl)dimethylsilyl]oxy]benzene, TRC

Product Code. 30351124
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30351124 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30351124 Supplier Toronto Research Chemicals Supplier No. TRCC369465100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(Chloromethyl)-3,5-bis(((1,1-dimethylethyl)dimethylsilyl)oxy)benzene
CAS 103929-85-7
Molecular Formula C19H35ClO2Si2
Quantity 100 mg
Synonym [[5-(Chloromethyl)-1,3-phenylene]bis(oxy)]bis[(1,1-dimethylethyl)dimethylsilane
InChI Formula InChI=1S/C19H35ClO2Si2/c1-18(2,3)23(7,8)21-16-11-15(14-20)12-17(13-16)22-24(9,10)19(4,5)6/h11-13H,14H2,1-10H3
SMILES ClCC1=CC(O[Si](C)(C)C(C)(C)C)=CC(O[Si](C)(C)C(C)(C)C)=C1
IUPAC Name tert-butyl-[3-[tert-butyl(dimethyl)silyl]oxy-5-(chloromethyl)phenoxy]-dimethylsilane
Molecular Weight (g/mol) 387.1
Formula Weight 386.19
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.