missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorohexane, 96%

Product Code. 11490477
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490477 50 g 50g
11400487 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11490477 Supplier Thermo Scientific Alfa Aesar Supplier No. B22379.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 544-10-5 | C6H13Cl | 120.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-10-5
Molecular Formula C6H13Cl
Molecular Weight (g/mol) 120.62
MDL Number MFCD00001018
InChI Key MLRVZFYXUZQSRU-UHFFFAOYSA-N
Synonym chlorohexane, hexyl chloride, hexane, 1-chloro, n-hexyl chloride, 1-chloro-hexane, hexane, chloro, unii-r5l7i6o9nw, r5l7i6o9nw, triisobutenyl chloride, hexylchloride
PubChem CID 10992
IUPAC Name 1-chlorohexane
SMILES CCCCCCCl

Specifications

Melting Point -94°C
Boiling Point 132°C to 134°C
Flash Point 26°C (78°F)
Linear Formula CH3(CH2)5Cl
Quantity 50 g
UN Number UN1993
Beilstein 1731289
Merck Index 14,2144
Formula Weight 120.62
Percent Purity 96%
Chemical Name or Material 1-Chlorohexane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.