missing translation for 'onlineSavingsMsg'
Learn More

1-Chloroethyl chloroformate, 98%

Product Code. 11349045
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11349045 25 g 25g
11339045 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11349045 Supplier Thermo Scientific Alfa Aesar Supplier No. L14197.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 50893-53-3 | C3H4Cl2O2 | 142.96 g/mol

1-Chloroethyl chloroformate acts as a reagent in the selective N-dealkylation of tertiary amines. It is also used as a reactant in the preparation of alkyl 1-chloroethyl carbonates. Further, it is used for N-demethylation reaction in the determination of multiple drugs of abuse in biological fluids using capillary electrophoresis with native fluorescence and laser-induced fluorescence detection. It plays an important role as a catalyst for the chemoselective desilylation of silyl-protected alcohols. In addition to this, it is used to cleave benzhydryl groups from amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloroethyl chloroformate acts as a reagent in the selective N-dealkylation of tertiary amines. It is also used as a reactant in the preparation of alkyl 1-chloroethyl carbonates. Further, it is used for N-demethylation reaction in the determination of multiple drugs of abuse in biological fluids using capillary electrophoresis with native fluorescence and laser-induced fluorescence detection. It plays an important role as a catalyst for the chemoselective desilylation of silyl-protected alcohols. In addition to this, it is used to cleave benzhydryl groups from amines.

Solubility
Miscible with most organic solvents.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agent, strong base, alcohols and amines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50893-53-3
Molecular Formula C3H4Cl2O2
Molecular Weight (g/mol) 142.96
MDL Number MFCD00000647
InChI Key QOPVNWQGBQYBBP-UHFFFAOYNA-N
Synonym 1-chloroethyl chloroformate, chloroformic acid 1-chloroethyl ester, 1-chloro ethyl chloroformate, 1-chloroethylchloroformate, carbonochloridic acid, 1-chloroethyl ester, ace-cl, alpha-chloroethyl chloroformate, chloro 1-chloroethoxy methanone, .alpha.-chloroethyl chloroformate, pubchem17418
PubChem CID 521305
IUPAC Name 1-chloroethyl carbonochloridate
SMILES CC(Cl)OC(Cl)=O

Specifications

Melting Point -65°C
Density 1.312
Boiling Point 119°C to 121°C
Flash Point 45°C (113°F)
Refractive Index 1.423
Quantity 25 g
UN Number UN2742
Beilstein 1747601
Sensitivity Moisture sensitive
Solubility Information Miscible with most organic solvents.
Formula Weight 142.97
Percent Purity 98%
Chemical Name or Material 1-Chloroethyl chloroformate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.