missing translation for 'onlineSavingsMsg'
Learn More

1-Chloroethyl chloroformate, 97+%

Product Code. 10397021 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10397021 100 g Glass bottle 100g
10727392 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10397021 Supplier Thermo Scientific Acros Supplier No. 314461000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 50893-53-3 | C3H4Cl2O2 | 142.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50893-53-3
Molecular Formula C3H4Cl2O2
Molecular Weight (g/mol) 142.96
MDL Number MFCD00000647
InChI Key QOPVNWQGBQYBBP-UHFFFAOYNA-N
Synonym 1-chloroethyl chloroformate, chloroformic acid 1-chloroethyl ester, 1-chloro ethyl chloroformate, 1-chloroethylchloroformate, carbonochloridic acid, 1-chloroethyl ester, ace-cl, alpha-chloroethyl chloroformate, chloro 1-chloroethoxy methanone, .alpha.-chloroethyl chloroformate, pubchem17418
PubChem CID 521305
IUPAC Name 1-chloroethyl carbonochloridate
SMILES CC(Cl)OC(Cl)=O

Specifications

CAS Min % 97.0
CAS Max % 100.0
Color Colorless
Density 1.3200g/mL
Boiling Point 118°C to 119°C
Flash Point 40°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Linear Formula ClCO2CH(Cl)CH3
Refractive Index 1.4220 to 1.424
Quantity 100 g
Specific Gravity 1.32
Formula Weight 142.97
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material 1-Chloroethyl chloroformate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.