missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorodecane, 97-99%

Product Code. p-86455 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
This item is not returnable. View return policy

Product Code. 10636102

Brand: Thermo Scientific Acros 108982500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1002-69-3 | C10H21Cl | 176.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1002-69-3
Molecular Formula C10H21Cl
Molecular Weight (g/mol) 176.73
MDL Number MFCD00000956
InChI Key ZTEHOZMYMCEYRM-UHFFFAOYSA-N
Synonym decyl chloride, decane, 1-chloro, n-decyl chloride, decane, chloro, unii-l0hsc44e5x, decyl chloride mixed isomers, chlorodecane, l0hsc44e5x, chlorodecane mixed isomers, 1-chlorodecane
PubChem CID 13848
IUPAC Name 1-chlorodecane
SMILES CCCCCCCCCCCl

Specifications

CAS Min % 97.0
CAS Max % 100.0
Melting Point -34°C
Color Colorless
Density 0.8600g/mL
Boiling Point 223°C
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)9Cl
Refractive Index 1.4362 to 1.4382
Quantity 250 mL
Beilstein 01, 168
Specific Gravity 0.86
Solubility Information Solubility in water: 0.01g/L (20°C)
Viscosity 1.94 mPa.s (20°C)
Formula Weight 176.73
Percent Purity 97-99%
Physical Form Liquid
Chemical Name or Material 1-Chlorodecane, 97-99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Chlorodecane
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.