missing translation for 'onlineSavingsMsg'
Learn More

(1-Chlorocyclohexyl)cyclohexyl-methanone, TRC

Product Code. 30366795
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30366795 100 mg 100mg
30351759 500 mg 500mg
30361462 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30366795 Supplier Toronto Research Chemicals Supplier No. TRCC371250100MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83803-75-2
Molecular Formula C13H21ClO
Molecular Weight (g/mol) 228.76
Synonym Ketone 1-Chlorocyclohexyl Cyclohexyl, (1-Chlorocyclohexyl)cyclohexylmethanone
IUPAC Name (1-chlorocyclohexyl)-cyclohexylmethanone
SMILES O=C(C1CCCCC1)C2(Cl)CCCCC2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C13H21ClO/c14-13(9-5-2-6-10-13)12(15)11-7-3-1-4-8-11/h11H,1-10H2
Formula Weight 228.1281
Chemical Name or Material (1-Chlorocyclohexyl)cyclohexyl-methanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.