missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorobutane, HPLC Grade, 99.5+%

Product Code. 11333588
Change view
Click to view available options
Quantity:
250 mL
1 L
4 L
4 x 1 L
Unit Size:
1 L
250mL
4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11333588 4 L 4 L
11353588 250 mL 250mL
11323588 1 L 1 L
11343588 4 x 1 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11333588 Supplier Thermo Scientific Alfa Aesar Supplier No. 039198.K7
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Maximum level of impurities: Evaporation residue 1ppm, H2O 0.02% | CAS: 109-69-3 | C4H9Cl | 92.57 g/mol

1-Chlorobutane is used as intermediates and solvents. It is used for HPLC, spectrophotometry and environmental testing. It is used as alkylating reagent in organic and organometallic compound synthesis. It is also employed as an antihelmintic in veterinary medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-69-3
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.57
MDL Number MFCD00001009
InChI Key VFWCMGCRMGJXDK-UHFFFAOYSA-N
Synonym butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane
PubChem CID 8005
IUPAC Name 1-chlorobutane
SMILES CCCCCl

Specifications

Melting Point °C to 123°C
Color Colorless
Density 0.886 g/cm3 at 20°C
Boiling Point 77°C to 78°C
Flash Point -12°C (10°F)
Ignition Point 280°C (536°F)
Assay Percent Range >99.5%
Odor Pungent
Refractive Index 1.4024
Quantity 4 L
Beilstein 1730909
Solubility Information Miscible with alcohol and ether. Immiscible with water.
Formula Weight 92.57
Vapor Pressure 110 hPa (83mm Hg) at 20°C
Grade HPLC
Concentration or Composition (by Analyte or Components) 1-Chlorobutane
Physical Form Liquid
Chemical Name or Material 1-Chlorobutane, Filtered through 0.2μ filters.
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.