missing translation for 'onlineSavingsMsg'
Learn More

1-Chloroanthraquinone, 98%, Thermo Scientific™

Product Code. p-84859 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
This item is not returnable. View return policy

Product Code. 10122470

Brand: Thermo Scientific Acros 108600050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 82-44-0
MDL Number MFCD00001189
InChI Key BOCJQSFSGAZAPQ-UHFFFAOYSA-N
Synonym 1-chloroanthraquinone, 1-chloro anthraquinone, anthraquinone, 1-chloro, 1-chloro-9,10-anthraquinone, 9,10-anthracenedione, 1-chloro, 1-chloranthrachinon, 1-chloranthrachinon czech, alpha-chloroanthraquinone, alpha-monochloroanthraquinone, unii-96s5sqv15v
PubChem CID 6709
IUPAC Name 1-chloroanthracene-9,10-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=CC=C3)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 157°C to 162°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Molecular Formula C14H7ClO2
Quantity 5 g
Beilstein 07,787
Solubility Information Solubility in water: insoluble
Molecular Weight (g/mol) 242.66
Formula Weight 242.66
Percent Purity 98%
Physical Form Chunks and/or Powder
Chemical Name or Material 1-Chloroanthraquinone, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.