missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Chloroanthraquinone, 98%

Product Code. 11460348
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460348 100 g 100g
11470348 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11460348 Supplier Alfa Aesar™ Supplier No. B24522.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
It is used as a dyestuff intermediate.

Solubility
Soluble in hot toluene. (almost transparency)

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82-44-0
Molecular Formula C14H7ClO2
Molecular Weight (g/mol) 242.658
MDL Number MFCD00001189
InChI Key BOCJQSFSGAZAPQ-UHFFFAOYSA-N
Synonym 1-chloroanthraquinone, 1-chloro anthraquinone, anthraquinone, 1-chloro, 1-chloro-9,10-anthraquinone, 9,10-anthracenedione, 1-chloro, 1-chloranthrachinon, 1-chloranthrachinon czech, alpha-chloroanthraquinone, alpha-monochloroanthraquinone, unii-96s5sqv15v
PubChem CID 6709
IUPAC Name 1-chloroanthracene-9,10-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=CC=C3)Cl

Specifications

Melting Point 158°C to 162°C
Quantity 100 g
Beilstein 1912752
Formula Weight 242.66
Percent Purity 98%
Chemical Name or Material 1-Chloroanthraquinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.