missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(Chloroacetyl)-2-(trifluoroacetyl)hydrazine, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 1-(Chloroacetyl)-2-(trifluoroacetyl)hydrazine |
| CAS | 762240-99-3 |
| Molecular Formula | C4 H4 Cl F3 N2 O2 |
| Quantity | 1 g |
| Synonym | Acetic acid, 2,2,2-trifluoro-, 2-(2-chloroacetyl)hydrazide (ACI), 2,2,2-Trifluoroacetic acid 2-(2-chloroacetyl)hydrazide (ACI), Acetic acid, trifluoro-, 2-(chloroacetyl)hydrazide (9CI), 1-(Chloroacetyl)-2-(trifluoroacetyl)hydrazine |
| InChI Formula | InChI=1S/C4H4ClF3N2O2/c5-1-2(11)9-10-3(12)4(6,7)8/h1H2,(H,9,11)(H,10,12) |
| SMILES | FC(F)(F)C(=O)NNC(=O)CCl |
| IUPAC Name | N'-(2-chloroacetyl)-2,2,2-trifluoroacetohydrazide |
| Molecular Weight (g/mol) | 204.54 |
| Formula Weight | 203.9913 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction