missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-N,N,2-trimethylpropenylamine, 98.5+%

Product Code. 10217750 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
This item is not returnable. View return policy

Product Code. 10217750

Brand: Thermo Scientific Acros 300270050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26189-59-3 | C6H12ClN | 133.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26189-59-3
Molecular Formula C6H12ClN
Molecular Weight (g/mol) 133.62
InChI Key GQIRIWDEZSKOCN-UHFFFAOYSA-N
Synonym 1-chloro-n,n,2-trimethylpropenylamine, 1-chloro-n,n,2-trimethyl-1-propenylamine, 1-chloro-n,n,2-trimethyl-1-propen-1-amine, ghosez inverted exclamation markas reagent, 1-propen-1-amine, 1-chloro-n,n,2-trimethyl, 1-chloro-2-methylprop-1-en-1-yl dimethylamine, acmc-1ceik, 1-chloro-1-dimethylaminoisobutene, 1-chloro-2,n,n-trimethylpropenylamine, 1-chloro-n,n-2-trimethylpropenylamine
PubChem CID 640792
IUPAC Name 1-chloro-N,N,2-trimethylprop-1-en-1-amine
SMILES CC(=C(N(C)C)Cl)C

Specifications

Color Colorless to Yellow
Density 1.0100g/mL
Boiling Point 129.0°C to 130.0°C
Flash Point 27°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4520 to 1.4560
Quantity 5 mL
Specific Gravity 1.01
Solubility Information Solubility in water: .. Other solubilities: soluble in most organic solvents
Formula Weight 133.62
Percent Purity 98.5+%
Physical Form Liquid
Chemical Name or Material 1-Chloro-N, N, 2-trimethylpropenylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Chloro-N,N,2-trimethylpropenylamine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.