missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloro-N,N-2-trimethylpropenylamine (>90%), TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 1-Chloro-N,N-2-trimethylpropenylamine |
| CAS | 26189-59-3 |
| Molecular Formula | C6H12ClN |
| Quantity | 5 g |
| Synonym | 1-Chloro-N,N,2-trimethyl-1-propen-1-amine, (1-Chloro-2-methylprop-1-en-1-yl)dimethylamine, (1-Chloro-2-methylpropenyl)dimethylamine, 1-Chloro-1-(dimethylamino)-2-methylpropene, 1-Chloro-N,N,2-trimethyl-1-propenylamine, 1-Chloro-N,N,2-trimethylprop-1-en-1-amine, 1-Chloro-N,N-2-trimethylpropenylamine, 1-Chloro-N,N-dimethyl-2-methyl-1-propenylamine, 1-Chloro-N,N-dimethyl-2-methylprop-1-en-1-amine, Ghosez's reagent, N-(1-Chloro-2-methylprop-1-enyl)-N,N-dimethylamine |
| InChI Formula | InChI=1S/C6H12ClN/c1-5(2)6(7)8(3)4/h1-4H3 |
| SMILES | CN(C)C(=C(C)C)Cl |
| IUPAC Name | 1-chloro-N,N,2-trimethylprop-1-en-1-amine |
| Molecular Weight (g/mol) | 133.62 |
| Formula Weight | 133.0658 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction