missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-6-iodohexane, TRC

Product Code. 30367688
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30367688 5 g -
30353610 1 g 1g
30359499 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30367688 Supplier Toronto Research Chemicals Supplier No. TRCC3700655G
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34683-73-3
Molecular Formula C6H12ClI
Molecular Weight (g/mol) 246.52
Synonym 1-Iodo-6-chlorohexane
IUPAC Name 1-chloro-6-iodohexane
SMILES ClCCCCCCI

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H12ClI/c7-5-3-1-2-4-6-8/h1-6H2
Formula Weight 245.9672
Chemical Name or Material 1-Chloro-6-iodohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.