missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-6-hydroxyhexane, 95%

Product Code. 10010900 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10010900 100 g 100g
10112760 25 g 25g
10761424 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10010900 Supplier Thermo Scientific Acros Supplier No. 109281000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2009-83-8 | C6H13ClO | 136.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2009-83-8
Molecular Formula C6H13ClO
Molecular Weight (g/mol) 136.62
MDL Number MFCD00002984
InChI Key JNTPTNNCGDAGEJ-UHFFFAOYSA-N
Synonym 6-chloro-1-hexanol, 6-chlorohexanol, 1-chloro-6-hydroxyhexane, 1-hexanol, 6-chloro, hexamethylene chlorohydrin, 1-chloro-6-hexanol, omega-chlorohexanol, unii-t2hit6n5uf, 6-chloro-hexanol, t2hit6n5uf
PubChem CID 74828
IUPAC Name 6-chlorohexan-1-ol
SMILES C(CCCCl)CCO

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0300g/mL
Boiling Point 108.0°C to 112.0°C (14.0 mmHg)
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula Cl(CH2)6OH
Refractive Index 1.4547 to 1.4567
Quantity 100 g
Specific Gravity 1.03
Formula Weight 136.62
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1-chloro-6-hydroxyhexane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.