missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-5-trimethylsilyl-4-pentyne, 97%

Product Code. 15450158
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15450158 25 g 25g
15440158 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15450158 Supplier Thermo Scientific Alfa Aesar Supplier No. H53393.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 77113-48-5 | C8H15ClSi | 174.74 g/mol

1-Chloro-5-trimethylsilyl-4-pentyne is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-5-trimethylsilyl-4-pentyne is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77113-48-5
Molecular Formula C8H15ClSi
Molecular Weight (g/mol) 174.74
MDL Number MFCD00671353
InChI Key UPVHIBLXQNNITH-UHFFFAOYSA-N
Synonym 5-chloro-1-pentynyl trimethylsilane, 1-chloro-5-trimethylsilyl-4-pentyne, silane, 5-chloro-1-pentynyl trimethyl, acmc-1bffc, 5-chloro-1-trimethylsilylpent-1-yne, 5-chloropent-1-ynyl trimethyl silane, 5-chloropent-1-yn-1-yl trimethylsilane, silane, 5-chloro-1-pentyn-1-yl trimethyl
PubChem CID 2757942
SMILES C[Si](C)(C)C#CCCCCl

Specifications

Boiling Point 75°C to 77°C (21mmHg)
Quantity 25 g
Linear Formula Cl(CH2)3C≡CSi(CH3)3
Flash Point 65°C (149°F)
IUPAC Name 5-chloropent-1-ynyl(trimethyl)silane
Formula Weight 174.75
Percent Purity 97%
Refractive Index 1.456
Density 0.977
Chemical Name or Material 1-Chloro-5-trimethylsilyl-4-pentyne

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.