missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-5-triethylsilyl-4-pentyne, 97%

Product Code. 15490418
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490418 5 g 5g
15400428 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15490418 Supplier Thermo Scientific Alfa Aesar Supplier No. H53515.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 174125-30-5 | C11H21ClSi | 216.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 174125-30-5
Molecular Formula C11H21ClSi
Molecular Weight (g/mol) 216.82
MDL Number MFCD00671352
InChI Key YOFYXFCOGXKCGG-UHFFFAOYSA-N
Synonym 1-chloro-5-triethylsilyl-4-pentyne, 5-chloropent-1-ynyl triethyl silane, 5-chloropent-1-ynyl triethylsilane, 1-triethylsilyl-5-chloro-1-pentyne, 5-chloranylpent-1-ynyl triethyl silane, 5-chloropent-1-yn-1-yl triethylsilane, 5-chloropent-1-yn-1-yl triethyl silane, silane, 5-chloro-1-pentyn-1-yl triethyl
PubChem CID 2757930
IUPAC Name 5-chloropent-1-ynyl(triethyl)silane
SMILES CC[Si](CC)(CC)C#CCCCCl

Specifications

Quantity 5 g
Linear Formula Cl(CH2)3C≡CSi(CH2CH3)3
Formula Weight 216.83
Percent Purity 97%
Chemical Name or Material 1-Chloro-5-triethylsilyl-4-pentyne

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.