missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-4-phenylbutane, 97%

Product Code. 11439303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439303 1 g 1g
11449303 5 g 5g
11459303 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439303 Supplier Thermo Scientific Alfa Aesar Supplier No. A11753.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4830-93-7 | C10H13Cl | 168.664 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4830-93-7
Molecular Formula C10H13Cl
Molecular Weight (g/mol) 168.664
MDL Number MFCD00037117
InChI Key FLLZCZIHURYEQP-UHFFFAOYSA-N
Synonym 4-chlorobutyl benzene, 1-chloro-4-phenylbutane, 4-phenylbutyl chloride, 1-4-chlorobutyl benzene, benzene, 4-chlorobutyl, phenylbutylchloride, 1-chloro4-phenylbutane, acmc-209kcr, 4-phenyl-1-chlorobutane, 4-phenyl-n-butyl chloride
PubChem CID 78543
IUPAC Name 4-chlorobutylbenzene
SMILES C1=CC=C(C=C1)CCCCCl

Specifications

Density 1.026
Boiling Point 125°C to 128°C (20 mmHg)
Flash Point 104°C (219°F)
Refractive Index 1.518
Quantity 1 g
Beilstein 2042123
Formula Weight 168.67
Percent Purity 97%
Chemical Name or Material 1-Chloro-4-phenylbutane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.