missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-4-nitrobenzene, 99%

Product Code. p-89414 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
1 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
1kg
5g
This item is not returnable. View return policy

Product Code. 10356360

Brand: Thermo Scientific Acros 109630050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100-00-5 | C6H4ClNO2 | 157.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-00-5
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.56
MDL Number MFCD00007285
InChI Key CZGCEKJOLUNIFY-UHFFFAOYSA-N
Synonym 4-chloronitrobenzene, p-chloronitrobenzene, p-nitrochlorobenzene, benzene, 1-chloro-4-nitro, 4-nitrochlorobenzene, pncb, 4-chloro-1-nitrobenzene, p-nitrophenyl chloride, p-nitrochlorobenzol, p-nitroclorobenzene
PubChem CID 7474
ChEBI CHEBI:34399
IUPAC Name 1-chloro-4-nitrobenzene
SMILES C1=CC(=CC=C1[N+](=O)[O-])Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 81.0°C to 84.0°C
Color Yellow
Density 1.2980g/mL
Boiling Point 242.0°C
Flash Point 127°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula ClC6H4NO2
Quantity 5 g
Beilstein 05, 243
Merck Index 15, 2153
Specific Gravity 1.298
Solubility Information Solubility in water: insoluble.,soluble in organic solvents
Formula Weight 157.56
Percent Purity 99%
Physical Form Crystals or Flakes
Chemical Name or Material 1-Chloro-4-nitrobenzene, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Chloro-4-nitrobenzene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 2
  • Germ cell mutagenicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.