missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-4-nitrobenzene, 98+%

Product Code. 11459150
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
This item is not returnable. View return policy

Product Code. 11459150

Brand: Thermo Scientific Alfa Aesar A15396.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100-00-5 | C6H4ClNO2 | 157.553 g/mol

1-Chloro-4-nitrobenzene is an important intermediate used in the preparation of 2,4-dinitrochlorobenzene and 3,4-dichloronitrobenzene. It is used as a raw material in the preparation of 4-nitrodiphenylamine, 4-chloroaninline and (4-[(4-aminobenzene)sulfonyl]aniline). It serves as a common antioxidant, which is found in rubber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-4-nitrobenzene is an important intermediate used in the preparation of 2,4-dinitrochlorobenzene and 3,4-dichloronitrobenzene. It is used as a raw material in the preparation of 4-nitrodiphenylamine, 4-chloroaninline and (4-[(4-aminobenzene)sulfonyl]aniline). It serves as a common antioxidant, which is found in rubber.

Solubility
Soluble in boiling hot ethanol, ether, toluene, acetone, organic solvents and carbon disulfide. Sparingly soluble in cold alcohol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-00-5
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.553
MDL Number MFCD00007285
InChI Key CZGCEKJOLUNIFY-UHFFFAOYSA-N
Synonym 4-chloronitrobenzene, p-chloronitrobenzene, p-nitrochlorobenzene, benzene, 1-chloro-4-nitro, 4-nitrochlorobenzene, pncb, 4-chloro-1-nitrobenzene, p-nitrophenyl chloride, p-nitrochlorobenzol, p-nitroclorobenzene
PubChem CID 7474
ChEBI CHEBI:34399
IUPAC Name 1-chloro-4-nitrobenzene
SMILES C1=CC(=CC=C1[N+](=O)[O-])Cl

Specifications

Melting Point 82°C to 86°C
Density 1.515
Boiling Point 238°C to 242°C
Flash Point 127°C (260°F)
Quantity 1000 g
UN Number UN1578
Beilstein 508691
Merck Index 14,2151
Solubility Information Soluble in boiling hot ethanol,ether,toluene,acetone,organic solvents and carbon disulfide. Sparingly soluble in cold alcohol. Insoluble in water.
Formula Weight 157.56
Percent Purity ≥98%
Chemical Name or Material 1-Chloro-4-nitrobenzene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Chloro-4-nitrobenzene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 2
  • Germ cell mutagenicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : 1-Chloro-4-nitrobenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.