missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-4-hydroxyisoquinoline, 97%

Product Code. 10432044 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10432044 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10432044 Supplier Thermo Scientific Acros Supplier No. 399780010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3336-43-4 | C9H6ClNO | 179.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3336-43-4
Melting Point 195°C to 197°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C9H6ClNO
MDL Number MFCD03425941
Quantity 1 g
Synonym 1-chloro-4-hydroxyisoquinoline, 4-isoquinolinol, 1-chloro, pubchem18457, 1-chloroisoquinoline-4-ol, 1-chloro-isoquinolin-4-ol, 4-isoquinolinol,1-chloro, 1-chloro-4-hydroxy isoquinoline, 1-chloro-4-hydroxyisoquinoline 97
InChI Key JEVLGPVFFYUBRI-UHFFFAOYSA-N
SMILES OC1=CN=C(Cl)C2=CC=CC=C12
IUPAC Name 1-chloroisoquinolin-4-ol
Molecular Weight (g/mol) 179.60
PubChem CID 4281882
Formula Weight 179.61
Percent Purity 97%
Chemical Name or Material 1-Chloro-4-hydroxyisoquinoline
Show More Show Less
compliance-icons
Product Identifier
  • 1-Chloro-4-hydroxyisoquinoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.