missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-4-ethynylbenzene, 98%

Product Code. 10778183 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10778183 1 g Glass bottle 1g
10536553 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10778183 Supplier Thermo Scientific Acros Supplier No. 377060010
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 873-73-4 | C8H5Cl | 136.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873-73-4
Molecular Formula C8H5Cl
Molecular Weight (g/mol) 136.58
MDL Number MFCD00191917
InChI Key LFZJRTMTKGYJRS-UHFFFAOYSA-N
Synonym 4-chlorophenylacetylene, benzene, 1-chloro-4-ethynyl, 4'-chlorophenyl acetylene, 4-chlorophenyl acetylene, 1-chloro-4-ethynyl-benzene, p-ethynylchlorobenzene, p-chlorophenylacetylene, p-chlorophenyl acetylene, 1-chlor-4-ethinylbenzol, 4-chlorophenylethyne
PubChem CID 70118
IUPAC Name 1-chloro-4-ethynylbenzene
SMILES C#CC1=CC=C(C=C1)Cl

Specifications

Melting Point 43°C to 47°C
CAS Min % 97.5
Color Brown to Yellow
CAS Max % 100.0
Quantity 1 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Mass
Assay Percent Range 97.5% min. (GC)
Linear Formula ClC6H4C∽CH
Beilstein 05,III,135
Boiling Point 79°C to 82°C (23.0 mmHg)
Formula Weight 136.58
Percent Purity 98%
Chemical Name or Material 1-Chloro-4-ethynylbenzene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Chloro-4-ethynylbenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.