missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Chloro-4-(4-fluorophenyl)phthalazine, 96%

Product Code. 15477417
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15477417

Brand: Alfa Aesar™ H34466.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 155937-28-3
Molecular Formula C14H8ClFN2
Molecular Weight (g/mol) 258.68
MDL Number MFCD06253841
InChI Key LIFADULWFCVAQR-UHFFFAOYSA-N
Synonym 1-chloro-4-4-fluorophenyl phthalazine, 4-chloro-1-4-fluorophenyl phthalazine, 1-chloro-4-4-fluorophenyl-phthalazine, 1-chloro-4-4-fluoro-phenyl-phthalazine, 1-chloro-4-4-fluorophenyl phthalazine im ocs
PubChem CID 2115239
IUPAC Name 1-chloro-4-(4-fluorophenyl)phthalazine
SMILES FC1=CC=C(C=C1)C1=NN=C(Cl)C2=CC=CC=C12

Specifications

Melting Point 196°C to 197°C
Quantity 250 mg
Formula Weight 258.68
Percent Purity 96%
Chemical Name or Material 1-Chloro-4-(4-fluorophenyl)phthalazine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.