missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3-methyl-2-butene, 95%, stab. with potassium carbonate

Product Code. 11457867
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11457867 50 g 50g
11447867 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11457867 Supplier Thermo Scientific Alfa Aesar Supplier No. A16545.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 503-60-6 | C5H9Cl | 104.577 g/mol

1-Chloro-3-methyl-2-butene is used in the synthesis of geraniol. It is also used as an alkylating agent and as a reagent in hyperforin. Further, it is used as an antibiotic to inhibit growth of tumor cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-3-methyl-2-butene is used in the synthesis of geraniol. It is also used as an alkylating agent and as a reagent in hyperforin. Further, it is used as an antibiotic to inhibit growth of tumor cells.

Solubility
Miscible with chloroform, acetone, diethyl ether and alcohol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 503-60-6
Molecular Formula C5H9Cl
Molecular Weight (g/mol) 104.577
MDL Number MFCD00000982
InChI Key JKXQKGNGJVZKFA-UHFFFAOYSA-N
Synonym 1-chloro-3-methyl-2-butene, prenyl chloride, 2-butene, 1-chloro-3-methyl, 3,3-dimethylallyl chloride, 3-methyl-2-butenyl chloride, 3-methylcrotyl chloride, unii-kmt7v9llbr, gamma,gamma-dimethylallyl chloride, kmt7v9llbr, .gamma.,.gamma.-dimethylallyl chloride
PubChem CID 10429
IUPAC Name 1-chloro-3-methylbut-2-ene
SMILES CC(=CCCl)C

Specifications

Density 0.928
Boiling Point 107°C to 109°C
Flash Point 13°C (56°F)
Linear Formula (CH3)2C=CHCH2Cl
Refractive Index 1.449
Quantity 50 g
UN Number UN1993
Solubility Information Miscible with chloroform,acetone,diethyl ether and alcohol. Immiscible with water.
Formula Weight 104.57
Percent Purity 95%
Chemical Name or Material 1-Chloro-3-methyl-2-butene, stabilized with potassium carbonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.