missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloro-3-methyl-2-butene (>85%), TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | 1-Chloro-3-methyl-2-butene |
| CAS | 503-60-6 |
| Molecular Formula | C5H9Cl |
| Quantity | 25 g |
| Synonym | 1,1-Dimethyl-3-chloro-1-propene, 2-Methyl-4-chloro-2-butene, 3,3-Dimethylallyl Chloride, 3-Methyl-1-chloro-2-butene, 3-Methyl-2-butenyl Chloride, 3-Methylcrotyl Chloride, 4-Chloro-2-methyl-2-butene, Isoprenyl Chloride, Prenyl Chloride, γ,γ-Dimethylallyl Chloride |
| InChI Formula | InChI=1S/C5H9Cl/c1-5(2)3-4-6/h3H,4H2,1-2H3 |
| SMILES | CC(=CCCl)C |
| IUPAC Name | 1-chloro-3-methylbut-2-ene |
| Molecular Weight (g/mol) | 104.58 |
| Formula Weight | 104.0393 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction