missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3-iodobenzene, 98%, Thermo Scientific™

Product Code. 10147883 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10147883 100 g Glass bottle 100g
10751881 5 g Glass bottle 5g
10124850 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10147883 Supplier Thermo Scientific Acros Supplier No. 221911000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 625-99-0
Molecular Formula C6H4ClI
Molecular Weight (g/mol) 238.45
MDL Number MFCD00001046
InChI Key JMLWXCJXOYDXRN-UHFFFAOYSA-N
Synonym 3-chloroiodobenzene, m-chloroiodobenzene, benzene, 1-chloro-3-iodo, 3-iodochlorobenzene, 3-chloro-1-iodobenzene, 1-chloro-3-iodo-benzene, m-chlorophenyl iodide, m-iodochlorobenzene, metachloroiodobenzene, 3-chloro-iodobenzene
PubChem CID 12269
IUPAC Name 1-chloro-3-iodobenzene
SMILES ClC1=CC=CC(I)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.9200g/mL
Boiling Point 103.0°C to 104.0°C (15.0 mmHg)
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula ClC6H4I
Refractive Index 1.6300 to 1.6320
Quantity 100 g
Beilstein 05, 220
Formula Weight 238.45
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-chloro-3-iodobenzene
Show More Show Less
compliance-icons
Product Identifier
  • 1-Chloro-3-iodobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.