missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3-iodobenzene, 98%, Thermo Scientific™

Catalog Number p-3757235
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
625-99-0
C6H4ClI
238.45
MFCD00001046
JMLWXCJXOYDXRN-UHFFFAOYSA-N
3-chloroiodobenzene, m-chloroiodobenzene, benzene, 1-chloro-3-iodo, 3-iodochlorobenzene, 3-chloro-1-iodobenzene, 1-chloro-3-iodo-benzene, m-chlorophenyl iodide, m-iodochlorobenzene, metachloroiodobenzene, 3-chloro-iodobenzene
12269
1-chloro-3-iodobenzene
ClC1=CC=CC(I)=C1
This item is not returnable. View return policy
625-99-0
C6H4ClI
238.45
MFCD00001046
JMLWXCJXOYDXRN-UHFFFAOYSA-N
3-chloroiodobenzene, m-chloroiodobenzene, benzene, 1-chloro-3-iodo, 3-iodochlorobenzene, 3-chloro-1-iodobenzene, 1-chloro-3-iodo-benzene, m-chlorophenyl iodide, m-iodochlorobenzene, metachloroiodobenzene, 3-chloro-iodobenzene
12269
1-chloro-3-iodobenzene
ClC1=CC=CC(I)=C1
CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.9200g/mL
Boiling Point 103.0°C to 104.0°C (15.0 mmHg)
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula ClC6H4I
Refractive Index 1.6300 to 1.6320
Quantity 5 g
Beilstein 05, 220
Formula Weight 238.45
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-chloro-3-iodobenzene
Show More Show Less
compliance-icons
Product Identifier
  • 1-Chloro-3-iodobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'