missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3,3-dimethylbutane, 97%

Product Code. 11372607
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11372607 50 g 50g
11362607 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372607 Supplier Thermo Scientific Alfa Aesar Supplier No. L11753.18
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2855-08-5 | C6H13Cl | 120.62 g/mol

1-Chloro-3,3-dimethylbutane is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-3,3-dimethylbutane is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Not miscible or difficult to mix in water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2855-08-5
MDL Number MFCD00018980

Specifications

Density 0.867
Boiling Point 116°C to 118°C
Flash Point 7°C (44°F)
Molecular Formula C14H12N4O4
Linear Formula (CH3)3CCH2CH2Cl
Refractive Index 1.417
Quantity 50 g
UN Number UN1993
Beilstein 1731465
Synonym m-tolualdehyde 2,4-dinitrophenylhydrazone, m-tolualdehyde, 2,4-dinitrophenyl hydrazone, benzaldehyde, 3-methyl-, 2,4-dinitrophenyl hydrazone, 3'-methylbenzaldehyde 2,4-dinitrophenylhydrazone, m-tolualdehyd-2,4-dinitrophenylhydrazone, m-tolualdehyde 2,4-dinitro-phenylhydrazone, 3-methylbenzaldehyde 2,4-dinitrophenyl hydrazone #, 1-2,4-dinitrophenyl-2-3-methylbenzylidene hydrazine, n-e-3-methylphenyl methylideneamino-2,4-dinitroaniline, m-tolualdehyde 2,4-dinitrophenylhydrazone, environmental standard
Solubility Information Not miscible or difficult to mix in water.
InChI Key PQBRXQUGVASQBU-UHFFFAOYSA-N
SMILES CC1=CC=CC(=C1)C=NNC2=C(C=C(C=C2)[N+](=O)[O-])[N+](=O)[O-]
IUPAC Name N-[(E)-(3-methylphenyl)methylideneamino]-2,4-dinitroaniline
Molecular Weight (g/mol) 300.27
PubChem CID 9604108
Formula Weight 120.62
Percent Purity 97%
Chemical Name or Material 1-Chloro-3,3-dimethylbutane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.