missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-octyne, 98%, Thermo Scientific Chemicals

Product Code. 15450278
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15450278 5 g 5g
15460278 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15450278 Supplier Thermo Scientific Alfa Aesar Supplier No. H53452.06
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 51575-83-8 | Molecular Formula: C8H13Cl | Molecular Weight: 144.642 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51575-83-8
Molecular Formula C8H13Cl
Molecular Weight (g/mol) 144.642
MDL Number MFCD00274223
InChI Key OUMUOQQKEAGFCJ-UHFFFAOYSA-N
Synonym 1-chloro-2-octyne, acmc-1asda, oumuoqqkeagfcj-uhfffaoysa
PubChem CID 4349444
IUPAC Name 1-chlorooct-2-yne
SMILES CCCCCC#CCCl

Specifications

Density 0.931
Boiling Point 81°C (15 mmHg)
Flash Point 68°C (154°F)
Linear Formula CH3(CH2)4C≡CCH2Cl
Refractive Index 1.46
Quantity 5 g
Formula Weight 144.64
Percent Purity 98%
Chemical Name or Material 1-Chloro-2-octyne

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.