missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-methyl-2-phenylpropane, 98%, Thermo Scientific™

Catalog Number p-88384
Click to view available options
Quantity:
5 mL
100 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
515-40-2
C10H13Cl
168.66
MFCD00000940
DNXXUUPUQXSUFH-UHFFFAOYSA-N
neophyl chloride, 1-chloro-2-methyl-2-phenylpropane, 1-chloro-2-methylpropan-2-yl benzene, benzene, 2-chloro-1,1-dimethylethyl, 2-methyl-2-phenylpropyl chloride, neophylchloride, 2-chloro-1,1-dimethylethyl benzene, unii-of6e0410nf, beta-chloro-tert-butylbenzene, 2-chloromethyl-2-phenylpropane
68191
(1-chloro-2-methylpropan-2-yl)benzene
CC(C)(CCl)C1=CC=CC=C1
This item is not returnable. View return policy
515-40-2
C10H13Cl
168.66
MFCD00000940
DNXXUUPUQXSUFH-UHFFFAOYSA-N
neophyl chloride, 1-chloro-2-methyl-2-phenylpropane, 1-chloro-2-methylpropan-2-yl benzene, benzene, 2-chloro-1,1-dimethylethyl, 2-methyl-2-phenylpropyl chloride, neophylchloride, 2-chloro-1,1-dimethylethyl benzene, unii-of6e0410nf, beta-chloro-tert-butylbenzene, 2-chloromethyl-2-phenylpropane
68191
(1-chloro-2-methylpropan-2-yl)benzene
CC(C)(CCl)C1=CC=CC=C1
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0400g/mL
Boiling Point 95°C to 100°C (10.0 mmHg)
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C(CH3)2CH2Cl
Refractive Index 1.5230 to 1.525
Quantity 5 mL
Beilstein 05,II,32
Merck Index 15,6544
Specific Gravity 1.04
Solubility Information Solubility in water: insoluble
Formula Weight 168.67
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Chloro-2-methyl-2-phenylpropane, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Chloro-2-methyl-2-phenylpropane
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'