missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-iodotetrafluoroethane, 97%

Product Code. 11379155
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379155 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11379155 Supplier Thermo Scientific Alfa Aesar Supplier No. L16430.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 421-78-3 | C2ClF4I | 262.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 421-78-3
Molecular Formula C2ClF4I
Molecular Weight (g/mol) 262.37
MDL Number MFCD00069092
InChI Key XNIBSTIKPHNKSO-UHFFFAOYSA-N
Synonym 1-chloro-2-iodotetrafluoroethane, 2-chlorotetrafluoroethyl iodide, acmc-1al8i, 1-iodo-2-chlorotetrafluoroethane, 1-iodo-2-chloro-1,1,2,2-tetrafluoroethane, 1-chloro-1,1,2,2-tetrafluoro-2-iodo-ethane, ethane,1-chloro-1,1,2,2-tetrafluoro-2-iodo, 1-chloranyl-1,1,2,2-tetrakis fluoranyl-2-iodanyl-ethane
PubChem CID 136268
IUPAC Name 1-chloro-1,1,2,2-tetrafluoro-2-iodoethane
SMILES C(C(F)(F)I)(F)(F)Cl

Specifications

Boiling Point 56°C
Refractive Index 1.392
Quantity 25 g
UN Number UN2810
Beilstein 1699458
Sensitivity Light sensitive
Formula Weight 262.37
Percent Purity 97%
Chemical Name or Material 1-Chloro-2-iodotetrafluoroethane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.