missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-iodoethane, 96%, stab. with copper

Product Code. 11302976
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11302976 1 g 1g
11312976 5 g 5g
11322976 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11302976 Supplier Thermo Scientific Alfa Aesar Supplier No. L10742.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 624-70-4 | C2H4ClI | 190.41 g/mol

1-Chloro-2-iodoethane is used as a mild electrophile for iodination of metallated species, as in the conversion of an ortho-lithiated benzyl ether to the corresponding ortho-iodide. It reacts with 5-hydroxy-[1,4]naphthoquinone, and obtain the 5-(2-chloro-ethoxy)-[1,4]naphthoquinone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-2-iodoethane is used as a mild electrophile for iodination of metallated species, as in the conversion of an ortho-lithiated benzyl ether to the corresponding ortho-iodide. It reacts with 5-hydroxy-[1,4]naphthoquinone, and obtain the 5-(2-chloro-ethoxy)-[1,4]naphthoquinone.

Solubility
Soluble inorganic solvents. Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-70-4
Molecular Formula C2H4ClI
Molecular Weight (g/mol) 190.41
MDL Number MFCD00041453
InChI Key JTWWWQGSFTWWDL-UHFFFAOYSA-N
Synonym 1,2-chloroiodoethane, 1-chloro-2-iodo-ethane, ethane, 1-chloro-2-iodo, 2-chloroethyl iodide, 2-iodoethyl chloride, 1-iodo-2-chloroethane, 2-chlor-1-jod-athan, 2-chloro-1-iodoethane, 2-chloro-1-iodo-ethane, acmc-1b3nb
PubChem CID 136438
IUPAC Name 1-chloro-2-iodoethane
SMILES ClCCI

Specifications

Density 2.088
Boiling Point 61°C to 62°C (30 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.566
Quantity 1 g
Beilstein 1730865
Sensitivity Light sensitive
Solubility Information Soluble inorganic solvents. Insoluble in water.
Formula Weight 190.41
Percent Purity 96%
Chemical Name or Material 1-Chloro-2-iodoethane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.