missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-iodobenzene, 99%, Thermo Scientific Chemicals

Product Code. 10766622
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-41-8
Molecular Formula C6H4ClI
Molecular Weight (g/mol) 238.45
MDL Number MFCD00001033
InChI Key MPEOPBCQHNWNFB-UHFFFAOYSA-N
Synonym 2-chloroiodobenzene, o-chloroiodobenzene, o-iodochlorobenzene, 2-iodochlorobenzene, benzene, 1-chloro-2-iodo, 2-chlorophenyl iodide, 2-chloro-1-iodobenzene, 1-chloro-2-iodo-benzene, chlorjodbenzol, iodochlorobenzene
PubChem CID 11993
IUPAC Name 1-chloro-2-iodobenzene
SMILES ClC1=CC=CC=C1I

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 1.0°C
Color Yellow
Density 1.9500g/mL
Boiling Point 234.0°C to 235.0°C
Flash Point 111°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula ClC6H4I
Refractive Index 1.6330 to 1.6350
Quantity 5 g
Beilstein 05, 220
Specific Gravity 1.95
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 238.46
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Chloro-2-iodobenzene, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Chloro-2-iodobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'