missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-cycloheptylethane, 96%, Thermo Scientific™

Catalog Number 11407224
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
84226-37-9
C9H17Cl
160.685
MFCD01632144
MPSIMZVBCYTZHR-UHFFFAOYSA-N
1-chloro-2-cycloheptylethane, 2-chloroethyl cycloheptane, 2-cycloheptylethyl chloride
20187818
2-chloroethylcycloheptane
C1CCCC(CC1)CCCl
This item is not returnable. View return policy
84226-37-9
C9H17Cl
160.685
MFCD01632144
MPSIMZVBCYTZHR-UHFFFAOYSA-N
1-chloro-2-cycloheptylethane, 2-chloroethyl cycloheptane, 2-cycloheptylethyl chloride
20187818
2-chloroethylcycloheptane
C1CCCC(CC1)CCCl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 5 g
Formula Weight 160.69
Percent Purity 96%
Chemical Name or Material 1-Chloro-2-cycloheptylethane
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'