missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2,2,2-trifluoroethyl difluoromethyl ether, 97%

Product Code. p-7058012
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.

Product Code. 11318386

Brand: Thermo Scientific Alfa Aesar L17315.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.

CAS: 26675-46-7 | C3H2ClF5O | 184.49 g/mol

1-Chloro-2,2,2-trifluoroethyl difluoromethyl ether is a useful solvent for dispersing fluorinated materials.

For Research and Development use only. Not to be used in clinical studies

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26675-46-7
Molecular Formula C3H2ClF5O
Molecular Weight (g/mol) 184.49
MDL Number MFCD00066609
InChI Key PIWKPBJCKXDKJR-UHFFFAOYNA-N
Synonym isoflurane, forane, 1-chloro-2,2,2-trifluoroethyl difluoromethyl ether, forene, aerrane, isoflurano, isofluranum, isoflo, compound 469, 2-chloro-2-difluoromethoxy-1,1,1-trifluoroethane
PubChem CID 3763
ChEBI CHEBI:6015
IUPAC Name 2-chloro-2-(difluoromethoxy)-1,1,1-trifluoroethane
SMILES FC(F)OC(Cl)C(F)(F)F

Specifications

Color Colorless
Density 1.496g/mL
Boiling Point 48°C to 49°C
Odor Ether-like
Refractive Index <1.3
Quantity 5 g
Beilstein 1852087
Merck Index 14,5175
Formula Weight 184.49
Percent Purity 97%
Chemical Name or Material 1-Chloro-2,2,2-trifluoroethyl difluoromethyl ether
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.