missing translation for 'onlineSavingsMsg'
Learn More

1-(chloro-1-pyrrolidinylmethylene)pyrrolidinium hexafluorophosphate, 98%, Thermo Scientific™

Product Code. 11976701 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11976701 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11976701 Supplier Thermo Scientific Acros Supplier No. 440620050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

 

Coupling reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 135540-11-3
CAS Min % 97.5
CAS Max % 100.0
Melting Point 152°C to 156°C
Color White
Elemental Analysis 31.68 to 33.31% C, 8.21 to 8.63% N
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C9H16ClF6N2P
MDL Number MFCD00191333
Quantity 5 g
Synonym pyclu, 1-chloro-1-pyrrolidinylmethylene pyrrolidinium hexafluorophosphate, chlorodipyrrolidinocarbenium hexafluorophosphate, 1-chloro pyrrolidin-1-yl methylene pyrrolidin-1-ium hexafluorophosphate v, 1-chloro-1-pyrrolidinylmethylene pyrrolidinium, pyciu, chlorodipyrrolidinocarbenium hexafluorophosphate, chloro-n,n,n',n'-bis tetramethylene formamidinium hexafluorophosphate, chloro-dipyrrolidinocarbenium hexafluorophosphate, pyclu chlorodipyrrolidinocarbenium hexafluorophosphate, 1-chloro-1-pyrrolidinylmethylene pyrrolidiniumhexafluorophosphate
InChI Key NHEGCUSBUWGOQM-UHFFFAOYSA-N
SMILES F[P-](F)(F)(F)(F)F.ClC(N1CCCC1)=[N+]1CCCC1
IUPAC Name 1-[chloro(pyrrolidin-1-ium-1-ylidene)methyl]pyrrolidine;hexafluorophosphate
Molecular Weight (g/mol) 332.66
PubChem CID 2736652
Formula Weight 332.66
Percent Purity 98%
Physical Form Powder or Solid
Chemical Name or Material 1-(chloro-1-pyrrolidinylmethylene)pyrrolidinium hexafluorophosphate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.