missing translation for 'onlineSavingsMsg'
Learn More

1-(Chloro-1-pyrrolidinylmethylene)pyrrolidinium hexafluorophosphate, 98+%, Thermo Scientific™

Product Code. 15440956
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15440956

Brand: Thermo Scientific Alfa Aesar H54247.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135540-11-3
Molecular Formula C9H16ClF6N2P
Molecular Weight (g/mol) 332.66
MDL Number MFCD00191333
InChI Key NHEGCUSBUWGOQM-UHFFFAOYSA-N
Synonym pyclu, 1-chloro-1-pyrrolidinylmethylene pyrrolidinium hexafluorophosphate, chlorodipyrrolidinocarbenium hexafluorophosphate, 1-chloro pyrrolidin-1-yl methylene pyrrolidin-1-ium hexafluorophosphate v, 1-chloro-1-pyrrolidinylmethylene pyrrolidinium, pyciu, chlorodipyrrolidinocarbenium hexafluorophosphate, chloro-n,n,n',n'-bis tetramethylene formamidinium hexafluorophosphate, chloro-dipyrrolidinocarbenium hexafluorophosphate, pyclu chlorodipyrrolidinocarbenium hexafluorophosphate, 1-chloro-1-pyrrolidinylmethylene pyrrolidiniumhexafluorophosphate
PubChem CID 2736652
IUPAC Name 1-[chloro(pyrrolidin-1-ium-1-ylidene)methyl]pyrrolidine;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.ClC(N1CCCC1)=[N+]1CCCC1

Specifications

Melting Point 153°C to 156°C
Quantity 25 g
Beilstein 7898575
Sensitivity Light and moisture sensitive
Formula Weight 332.65
Percent Purity ≥98%
Chemical Name or Material 1-(Chloro-1-pyrrolidinylmethylene)pyrrolidinium hexafluorophosphate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.