missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-1-phenylethane, 97%

Product Code. 10161131 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10161131 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10161131 Supplier Thermo Scientific Acros Supplier No. 327061000
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 672-65-1 | C8H9Cl | 140.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 672-65-1
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0600g/mL
Boiling Point 90°C (33.0 mmHg)
Flash Point 44°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C8H9Cl
Refractive Index 1.5260 to 1.528
MDL Number MFCD00018870
Quantity 100 g
Specific Gravity 1.06
Synonym 1-chloroethyl benzene, 1-chloro-1-phenylethane, 1-phenylethyl chloride, alpha-phenylethyl chloride, alpha-methylbenzyl chloride, benzene, 1-chloroethyl, 1-phenyl-ethylchloride, ccris 1778, .alpha.-methylbenzyl chloride, rs-phenethyl chloride
InChI Key GTLWADFFABIGAE-UHFFFAOYSA-N
SMILES CC(C1=CC=CC=C1)Cl
IUPAC Name 1-chloroethylbenzene
Molecular Weight (g/mol) 140.61
PubChem CID 12648
Formula Weight 140.61
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Chloro-1-phenylethane, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Chloro-1-phenylethane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.