missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Butylimidazole, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 1-Butylimidazole |
| CAS | 4316-42-1 |
| Molecular Formula | C7H12N2 |
| Quantity | 5 g |
| Synonym | 1-Butyl-1H-imidazole, 1-Butylimidazole, Butylimidazole, N-Butyl-1H-imidazole, N-Butylimidazole, NSC 158165 |
| InChI Formula | InChI=1S/C7H12N2/c1-2-3-5-9-6-4-8-7-9/h4,6-7H,2-3,5H2,1H3 |
| SMILES | CCCCn1ccnc1 |
| IUPAC Name | 1-butylimidazole |
| Molecular Weight (g/mol) | 124.18 |
| Formula Weight | 124.1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction