missing translation for 'onlineSavingsMsg'
Learn More

(1-Butyl)triphenylphosphonium bromide, 99%, Thermo Scientific™

Product Code. 11418753
Click to view available options
Quantity:
25 g
500 g
Unit Size:
25g
500g
This item is not returnable. View return policy

Product Code. 11418753

Brand: Thermo Scientific Alfa Aesar A10504.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1779-51-7
Molecular Formula C22H24BrP
Molecular Weight (g/mol) 399.31
MDL Number MFCD00011855
InChI Key IKWKJIWDLVYZIY-UHFFFAOYSA-M
Synonym butyltriphenylphosphonium bromide, n-butyltriphenylphosphonium bromide, 1-butyl triphenylphosphonium bromide, phosphonium, butyltriphenyl-, bromide, butyl triphenyl phosphonium bromide, butyl triphenyl phosphine bromide, n-butyltriphenylphosphonium chloride, butyltriphenylphosphanium bromide, phosphonium, butyltriphenyl-, bromide 1:1, n-butyl triphenylphosphonium bromide
PubChem CID 159628
IUPAC Name butyltriphenylphosphanium bromide
SMILES [Br-].CCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 240°C to 243°C
Quantity 25 g
UN Number UN3464
Beilstein 3580224
Formula Weight 399.32
Percent Purity 99%
Chemical Name or Material (1-Butyl)triphenylphosphonium bromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.