missing translation for 'onlineSavingsMsg'
Learn More

(1-Butyl)triethylammonium bromide, 97%

Product Code. 11471137
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11471137

Brand: Thermo Scientific Alfa Aesar B24128.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13028-69-8 | C10H24BrN | 238.213 g/mol

(1-Butyl)triethylammonium bromide is an important organic intermediate. It can be used in agrochemical, pharmaceutical & dye stuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Butyl)triethylammonium bromide is an important organic intermediate. It can be used in agrochemical, pharmaceutical & dye stuff field.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents, water/ moisture. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13028-69-8
Molecular Formula C10H24BrN
Molecular Weight (g/mol) 238.213
MDL Number MFCD00055541
InChI Key ZNFYPPCUDSCDPR-UHFFFAOYSA-M
Synonym 1-butyl triethylammonium bromide, butyltriethylammonium bromide, n,n,n-triethylbutan-1-aminium bromide, butyltriethylazanium bromide, acmc-20apri, triethylbutyl ammonium bromide, butyl-triethyl-ammonium bromide, triethyl-n-butylammonium bromide, 1-butanaminium,n,n,n-triethyl-, bromide 1:1
PubChem CID 11746670
IUPAC Name butyl(triethyl)azanium;bromide
SMILES CCCC[N+](CC)(CC)CC.[Br-]

Specifications

Melting Point 212°C to 215°C
Quantity 100 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 238.21
Percent Purity 97%
Chemical Name or Material (1-Butyl)triethylammonium bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.