missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl isothiocyanate, 98+%, Thermo Scientific Chemicals

Catalog Number 11444343
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
592-82-5
C5H9NS
115.19
MFCD00004824
LIMQQADUEULBSO-UHFFFAOYSA-N
butyl isothiocyanate, n-butyl isothiocyanate, butyl mustard oil, butane, 1-isothiocyanato, 1-butyl isothiocyanate, butylisothiocyanate, isothiocyanic acid, butyl ester, isothiocyanic acid n-butyl ester, 1-isothiocyanato-butane, butanisothiocyanate
11613
CHEBI:50534
1-isothiocyanatobutane
CCCCN=C=S
This item is not returnable. View return policy
592-82-5
C5H9NS
115.19
MFCD00004824
LIMQQADUEULBSO-UHFFFAOYSA-N
butyl isothiocyanate, n-butyl isothiocyanate, butyl mustard oil, butane, 1-isothiocyanato, 1-butyl isothiocyanate, butylisothiocyanate, isothiocyanic acid, butyl ester, isothiocyanic acid n-butyl ester, 1-isothiocyanato-butane, butanisothiocyanate
11613
CHEBI:50534
1-isothiocyanatobutane
CCCCN=C=S

1-Butyl isothiocyanate participates in the synthesis and characterization of Poly(N-Propargylthiourea). Participates in the Synthesis of 2-Aminobenzothiazoles via Copper (I)-Catalyzed Cross-Coupling with Part-Per-Million Catalyst Loadings. Thiols reactions with 1-Butyl isothiocyanate in aquatic toxicity data in the TETRATOX assay and reactivity data in an abiotic thiol assay. utyl isothiocyanate has been used as capping reagent to immobilize the functionalized derivative of Oregon Green 514 molecular switch on a preformed amino self assembled monlayer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Butyl isothiocyanate participates in the synthesis and characterization of Poly(N-Propargylthiourea). Participates in the Synthesis of 2-Aminobenzothiazoles via Copper (I)-Catalyzed Cross-Coupling with Part-Per-Million Catalyst Loadings. Thiols reactions with 1-Butyl isothiocyanate in aquatic toxicity data in the TETRATOX assay and reactivity data in an abiotic thiol assay. utyl isothiocyanate has been used as capping reagent to immobilize the functionalized derivative of Oregon Green 514 molecular switch on a preformed amino self assembled monlayer.

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture.
TRUSTED_SUSTAINABILITY
Density 0.954
Boiling Point 63°C to 65°C (11 mmHg)
Flash Point 66°C (150°F)
Odor Pungent
Linear Formula CH3(CH2)3NCS
Refractive Index 1.5
Quantity 100 g
UN Number UN2922
Beilstein 906839
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 115.2
Percent Purity ≥98%
Chemical Name or Material 1-Butyl isothiocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'