missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl isothiocyanate, 98+%

Product Code. 11434343
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434343 25 g 25g
11424343 5 g 5g
11444343 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11434343 Supplier Thermo Scientific Alfa Aesar Supplier No. A10935.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 592-82-5 | C5H9NS | 115.19 g/mol

1-Butyl isothiocyanate participates in the synthesis and characterization of Poly(N-Propargylthiourea). Participates in the Synthesis of 2-Aminobenzothiazoles via Copper (I)-Catalyzed Cross-Coupling with Part-Per-Million Catalyst Loadings. Thiols reactions with 1-Butyl isothiocyanate in aquatic toxicity data in the TETRATOX assay and reactivity data in an abiotic thiol assay. utyl isothiocyanate has been used as capping reagent to immobilize the functionalized derivative of Oregon Green 514 molecular switch on a preformed amino self assembled monlayer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Butyl isothiocyanate participates in the synthesis and characterization of Poly(N-Propargylthiourea). Participates in the Synthesis of 2-Aminobenzothiazoles via Copper (I)-Catalyzed Cross-Coupling with Part-Per-Million Catalyst Loadings. Thiols reactions with 1-Butyl isothiocyanate in aquatic toxicity data in the TETRATOX assay and reactivity data in an abiotic thiol assay. utyl isothiocyanate has been used as capping reagent to immobilize the functionalized derivative of Oregon Green 514 molecular switch on a preformed amino self assembled monlayer.

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-82-5
Molecular Formula C5H9NS
Molecular Weight (g/mol) 115.19
MDL Number MFCD00004824
InChI Key LIMQQADUEULBSO-UHFFFAOYSA-N
Synonym butyl isothiocyanate, n-butyl isothiocyanate, butyl mustard oil, butane, 1-isothiocyanato, 1-butyl isothiocyanate, butylisothiocyanate, isothiocyanic acid, butyl ester, isothiocyanic acid n-butyl ester, 1-isothiocyanato-butane, butanisothiocyanate
PubChem CID 11613
ChEBI CHEBI:50534
IUPAC Name 1-isothiocyanatobutane
SMILES CCCCN=C=S

Specifications

Density 0.954
Boiling Point 63°C to 65°C (11 mmHg)
Flash Point 66°C (150°F)
Odor Pungent
Linear Formula CH3(CH2)3NCS
Refractive Index 1.5
Quantity 25 g
UN Number UN2922
Beilstein 906839
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 115.2
Percent Purity ≥98%
Chemical Name or Material 1-Butyl isothiocyanate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Butyl isothiocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 1-Butyl isothiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.