Learn More
1-Butyl isothiocyanate, 98+%, Thermo Scientific Chemicals
Use promo code "21615" to get your promotional price.
Description
1-Butyl isothiocyanate participates in the synthesis and characterization of Poly(N-Propargylthiourea). Participates in the Synthesis of 2-Aminobenzothiazoles via Copper (I)-Catalyzed Cross-Coupling with Part-Per-Million Catalyst Loadings. Thiols reactions with 1-Butyl isothiocyanate in aquatic toxicity data in the TETRATOX assay and reactivity data in an abiotic thiol assay. utyl isothiocyanate has been used as capping reagent to immobilize the functionalized derivative of Oregon Green 514 molecular switch on a preformed amino self assembled monlayer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Butyl isothiocyanate participates in the synthesis and characterization of Poly(N-Propargylthiourea). Participates in the Synthesis of 2-Aminobenzothiazoles via Copper (I)-Catalyzed Cross-Coupling with Part-Per-Million Catalyst Loadings. Thiols reactions with 1-Butyl isothiocyanate in aquatic toxicity data in the TETRATOX assay and reactivity data in an abiotic thiol assay. utyl isothiocyanate has been used as capping reagent to immobilize the functionalized derivative of Oregon Green 514 molecular switch on a preformed amino self assembled monlayer.
Solubility
Insoluble in water.
Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture.
Specifications
Boiling Point | 63°C to 65°C (11 mmHg) |
Flash Point | 66°C (150°F) |
Odor | Pungent |
Linear Formula | CH3(CH2)3NCS |
Quantity | 5 g |
UN Number | UN2922 |
Beilstein | 906839 |
Formula Weight | 115.2 |
Percent Purity | ≥98% |
Chemical Name or Material | 1-Butyl isothiocyanate |
RUO – Research Use Only