missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Butyl-3-methylimidazolium Tetrafluoroborate, TRC
Click to view available options
Quantity:
250 g
Unit Size:
250g
Specifications
Specifications
| Chemical Name or Material | 1-Butyl-3-methylimidazolium Tetrafluoroborate |
| CAS | 174501-65-6 |
| Molecular Formula | C8H15N2 . (BF4) |
| Quantity | 250 g |
| Synonym | 1-Butyl-1-methylimidazolium Tetrafluoroborate, 1-Butyl-3-methyl-1H-imidazolium Tetrafluoroborate, 1-Butyl-3-methyl-1H-imidazolium Tetrafluoroborate(1-), 1-Butyl-3-methyl-1H-imidazolium Tetrafluoroborate(1-), 1-Butyl-3-methylimidazolium Tetrafluoroborate, 1-Butyl-3-methylimidazolium Tetrafluoroborate(1-), 1-Methyl-3-butylimidazolium Tetrafluoroborate, 1-Methyl-3-butylimidazolium Tetrafluoroborate(1-), 1-n-Butyl-3-methylimidazolium Tetrafluoroborate, 3-Butyl-1-methylimidazolium Tetrafluoroborate, BMIm Tetrafluoroborate(1-), Bmim Tetrafluoroborate, BmimBF4, Butylmethylimidazolium Tetrafluoroborate |
| InChI Formula | InChI=1S/C8H15N2.BF4/c1-3-4-5-10-7-6-9(2)8-10;2-1(3,4)5/h6-8H,3-5H2,1-2H3;/q+1;-1 |
| SMILES | CCCC[n+]1ccn(C)c1.F[B-](F)(F)F |
| IUPAC Name | 1-butyl-3-methylimidazol-3-ium;tetrafluoroborate |
| Molecular Weight (g/mol) | 226.03 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction