missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-3-methylimidazolium hexafluorophosphate, 98+%

Product Code. p-4258822 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10612364

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 174501-64-5 | C8H15F6N2P | 284.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ionic liquid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 174501-64-5
Molecular Formula C8H15F6N2P
Molecular Weight (g/mol) 284.19
MDL Number MFCD03093295
InChI Key IXQYBUDWDLYNMA-UHFFFAOYSA-N
Synonym 1-butyl-3-methylimidazolium hexafluorophosphate, 1-butyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, unii-zge3n4o8q9, 1-butyl-3-methylimidazoliumhexafluorophosphate, 1-n-butyl-3-methylimidazolium hexafluorophosphate, zge3n4o8q9, butylmethylimidazolium hexafluorophosphate, 1-methyl-3-butylimidazolium hexafluorophosphate, bmim pf6, c4mim hexafluorophosphate
PubChem CID 2734174
SMILES F[P-](F)(F)(F)(F)F.CCCCN1C=C[N+](C)=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 6.5°C
Color Colorless to Yellow
Density 1.3727g/mL
Boiling Point >340.0°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15,1584
Specific Gravity 1.3727
IUPAC Name 1-butyl-3-methylimidazol-3-ium;hexafluorophosphate
Viscosity 272.1 mPa.s (25°C)
Formula Weight 284.18
Percent Purity 98+%
Physical Form Viscous Liquid
Chemical Name or Material 1-Butyl-3-methylimidazolium hexafluorophosphate
Show More Show Less
compliance-icons
Product Identifier
  • 1-Butyl-3-methylimidazolium hexafluorophosphate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 1-Butyl-3-methylimidazolium hexafluorophosphate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.