missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-3-methylimidazolium chloride, 98+%

Product Code. 10490242 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
This item is not returnable. View return policy

Product Code. 10490242

Brand: Thermo Scientific Acros 354090250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 79917-90-1 | C8H15ClN2 | 174.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Ionic liquid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79917-90-1
Molecular Formula C8H15ClN2
Molecular Weight (g/mol) 174.67
MDL Number MFCD03095425
InChI Key FHDQNOXQSTVAIC-UHFFFAOYSA-M
Synonym 1-butyl-3-methylimidazolium chloride, 3-butyl-1-methyl-1h-imidazol-3-ium chloride, 1-n-butyl-3-methylimidazolium chloride, bmimcl, bmim cl, c4mim chloride, dsstox_cid_11461, dsstox_rid_78878, dsstox_gsid_31461
PubChem CID 2734161
IUPAC Name 1-butyl-3-methylimidazol-3-ium;chloride
SMILES [Cl-].CCCCN1C=C[N+](C)=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 73.0°C
Color Cream to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 1584
Solubility Information Solubility in water: soluble
Viscosity 170.1 mPa.s (80°C)
Formula Weight 174.67
Percent Purity 98+%
Physical Form Mass
Chemical Name or Material 1-Butyl-3-methylimidazolium chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Butyl-3-methylimidazolium chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.