missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-2,3-dimethylimidazolium hexafluorophosphate, 99%

Product Code. 11397399
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397399 50 g 50g
11387399 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397399 Supplier Thermo Scientific Alfa Aesar Supplier No. H27827.18
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 227617-70-1 | C9H17F6N2P | 298.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 227617-70-1
Molecular Formula C9H17F6N2P
Molecular Weight (g/mol) 298.21
MDL Number MFCD03790877
InChI Key JWFPQAXAGSAKRF-UHFFFAOYSA-N
Synonym 1-butyl-2,3-dimethylimidazolium hexafluorophosphate, 1-butyl-2,3-dimethyl-1h-imidazol-3-ium hexafluorophosphate v, 1-butyl-2,3-dimethylimidazol-1-ium hexafluorophosphate, 3-butyl-1,2-dimethylimidazol-1-ium hexafluorophosphate, acmc-209fyn, bdmim pf6, c4c1mim pf6, c4mmim pf6, dsstox_cid_27883, dsstox_rid_82636
PubChem CID 11243457
IUPAC Name 1-butyl-2,3-dimethylimidazol-3-ium;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.CCCCN1C=C[N+](C)=C1C

Specifications

Melting Point 48°C to 50°C
Density 1.354
Refractive Index 1.423
Quantity 50 g
Formula Weight 298.21
Percent Purity 99%
Chemical Name or Material 1-Butyl-2,3-dimethylimidazolium hexafluorophosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.