missing translation for 'onlineSavingsMsg'
Learn More

1-Butanol, HPLC Grade, 99%, Thermo Scientific Chemicals

Catalog Number 11378197
Click to view available options
Quantity:
1 L
4 x 1 L
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
71-36-3
C4H10O
74.123
MFCD00002964
LRHPLDYGYMQRHN-UHFFFAOYSA-N
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
263
CHEBI:28885
butan-1-ol
CCCCO
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
71-36-3
C4H10O
74.123
MFCD00002964
LRHPLDYGYMQRHN-UHFFFAOYSA-N
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
263
CHEBI:28885
butan-1-ol
CCCCO

Filtered through 0.2μm filters

1-Butanol is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point −90°C to -89°C
Density 0.810
Boiling Point 117°C to 118°C
Quantity 1 L
Flash Point 35°C (95°F)
Physical Form Liquid
Refractive Index 1.399
Assay Percent Range 99%
Beilstein 969148
Solubility Information Miscible with alcohols,ketones,aldehydes,ethers,aliphatic and aromatic hydrocarbons. Slightly miscible with water.
Formula Weight 74.12
Grade HPLC
Chemical Name or Material 1-Butanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'