missing translation for 'onlineSavingsMsg'
Learn More

1-Butanol, BAKER ANALYZED™ ACS, J.T. Baker™

Product Code. 10624771 Shop All J.T.Baker Products
Change view
Click to view available options
:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10624771 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10624771 Supplier JT Baker Supplier No. 8017.2500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Exceeds ACS Specifications

Specifications

Melting Point -89°C
CAS 71-36-3
Color Max. 10 (APHA)
Density 0.812 g/cm³ (20°C)
Quantity 2.5 L
Flash Point 30°C
Packaging Glass Bottle
Residue after Evaporation max. 0.001%
Assay Percent Range min. 99.4%
Titratable Acid max. 0.0008 meq/g
Molecular Formula C4H10O
MDL Number MFCD00002964
Water max. 0.1%
Boiling Point 117.6°C (1013 hPa)
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
SMILES CCCCO
IUPAC Name butan-1-ol
Molecular Weight (g/mol) 74.12
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.